• USA Home
  • Search Results

  • 3amino9ethylcarbazole2102713232111

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

3-Amino-9-ethylcarbazole

3 Products
Synonym:   9-Ethylcarbazol-3-amine, AEC
Image of 3-<WBR>Amino-<WBR>9-<WBR>ethylcarbazole
CAS Number:
Empirical Formula (Hill Notation):
 C14H14N2
Molecular Weight:
 210.27
 Beilstein/REAXYS Number:
 174969
 EC Number:
 MDL number:
 PubChem Substance ID:
Popular Documents:  Datasheet (PDF)  |  Specification Sheet (PDF)  |  FTNMR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Assay
  Physical Form
Show More