• USA Home
  • Search Results

  • 3aminobutanoicacid1031254148011

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

3-Aminobutanoic acid

1 Products
Synonym:   (±)-3-Aminobutyric acid, DL-3-Aminobutyric acid
Image of 3-Aminobutanoic acid
CAS Number:
Linear Formula:
 CH3CH(NH2)CH2COOH
Molecular Weight:
 103.12
 Beilstein/REAXYS Number:
 1720563
 EC Number:
 MDL number:
 NACRES:
 NA.22
 PubChem Substance ID:
Popular Documents:  FTNMR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Assay
  Physical Form
  Color
Show More

Product #
Description
A44207 3-Aminobutanoic acid, 97%,