• USA Home
  • Search Results

  • 3aminophenylboronicacidmonohydrate1549620665889111

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

3-Aminophenylboronic acid monohydrate

1 Products
Synonym:   3-Aminobenzeneboronic acid
Image of 3-Aminophenylboronic acid monohydrate
CAS Number:
Linear Formula:
 H2NC6H4B(OH)2 · H2O
Molecular Weight:
 154.96
 Beilstein/REAXYS Number:
 2936342
 MDL number:
 NACRES:
 NA.22
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)  |  RAMAN FTIR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More