• USA Home
  • Search Results

  • 3aminopyridine941146208811

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

3-Aminopyridine

1 Products
Synonym:   3-Pyridinamine
Image of 3-Aminopyridine
CAS Number:
Empirical Formula (Hill Notation):
 C5H6N2
Molecular Weight:
 94.11
 Beilstein/REAXYS Number:
 105692
 EC Number:
 MDL number:
 NACRES:
 NA.22
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)  |  FTNMR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More

Product #
Description
A78209 3-Aminopyridine, 99%,