• USA Home
  • Search Results

  • 3aminosalicylicacid1531457023011

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

3-Aminosalicylic acid

1 Products
Synonym:   3-Amino-2-hydroxybenzoic acid
Image of 3-Aminosalicylic acid
CAS Number:
Linear Formula:
 H2NC6H3-2-(OH)CO2H
Molecular Weight:
 153.14
 Beilstein/REAXYS Number:
 2090427
 EC Number:
 MDL number:
 NACRES:
 NA.22
 PubChem Substance ID:
Popular Documents:  FTNMR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More