• USA Home
  • Search Results

  • 3bromobenzylbromide2499382378911

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

3-Bromobenzyl bromide

1 Products
Synonym:   α,3-Dibromotoluene
Image of 3-Bromobenzyl bromide
CAS Number:
Linear Formula:
 BrC6H4CH2Br
Molecular Weight:
 249.93
 Beilstein/REAXYS Number:
 2078683
 EC Number:
 MDL number:
 NACRES:
 NA.22
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)  |  FTNMR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More