• USA Home
  • Search Results

  • 3bromoiodobenzene2829059118411

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

3-Bromoiodobenzene

1 Products
Image of 3-Bromoiodobenzene
CAS Number:
Linear Formula:
 BrC6H4I
Molecular Weight:
 282.90
 EC Number:
 MDL number:
 PubChem Substance ID:
Popular Documents:  FTNMR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More

Product #
Description
280097 3-Bromoiodobenzene, 98%,