• USA Home
  • Search Results

  • 3butyn1ol700992774211

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

3-Butyn-1-ol

1 Products
Image of 3-Butyn-1-ol
CAS Number:
Linear Formula:
 HC≡CCH2CH2OH
Molecular Weight:
 70.09
 Beilstein/REAXYS Number:
 773710
 EC Number:
 MDL number:
 NACRES:
 NA.22
 PubChem Substance ID:
 eCl@ss:
 39020335
Popular Documents:  Specification Sheet (PDF)  |  FTNMR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More

Product #
Description
130850 3-Butyn-1-ol, 97%,