• USA Home
  • Search Results

  • 3carboxyproxyl18623215468911

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

3-Carboxy-PROXYL

1 Products
Synonym:   3-(Carboxy)-2,2,5,5-tetramethyl-1-pyrrolidinyloxy, 3-Carboxy-2,2,5,5-tetramethyl-1-pyrrolidinyloxy, free radical
Image of 3-Carboxy-PROXYL
CAS Number:
Empirical Formula (Hill Notation):
 C9H16NO3
Molecular Weight:
 186.23
 EC Number:
 MDL number:
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)  |  FTIR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More

Product #
Description
253324 3-Carboxy-PROXYL, 98%,