• USA Home
  • Search Results

  • 3chloro12propanediol110549624211

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

(±)-3-Chloro-1,2-propanediol

3 Products
Synonym:   α-Chlorohydrin, α-Glycerol chlorohydrin, α-Monochlorohydrin, 3-MCPD
Image of (±)<WBR>-<WBR>3-<WBR>Chloro-<WBR>1,2-<WBR>propanediol
CAS Number:
Linear Formula:
 ClCH2CH(OH)CH2OH
Molecular Weight:
 110.54
 Beilstein/REAXYS Number:
 635684
 EC Number:
 MDL number:
 NACRES:
 NA.22
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Solubility
  Application(s)
  Assay
Show More