• USA Home
  • Search Results

  • 3chloro1phenyl1propyne15060335531511

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

3-Chloro-1-phenyl-1-propyne

1 Products
Synonym:   1-Chloro-3-phenyl-2-propyne, 3-Phenylpropargyl chloride
Image of 3-<WBR>Chloro-<WBR>1-<WBR>phenyl-<WBR>1-<WBR>propyne
CAS Number:
Empirical Formula (Hill Notation):
 C9H7Cl
Molecular Weight:
 150.60
 MDL number:
 NACRES:
 NA.22
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More