• USA Home
  • Search Results

  • 3chloro2fluoroaniline14556210604911

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

3-Chloro-2-fluoroaniline

1 Products
Image of 3-<WBR>Chloro-<WBR>2-<WBR>fluoroaniline
CAS Number:
Linear Formula:
 ClC6H3(F)NH2
Molecular Weight:
 145.56
 EC Number:
 MDL number:
 NACRES:
 NA.22
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More