• USA Home
  • Search Results

  • 3chloro4morpholin4yl125thiadiazole205673016596911

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

3-Chloro-4-morpholin-4-yl-1,2,5-thiadiazole

1 Products
Synonym:   4-(4-Chloro-1,2,5-thiadiazol-3-yl)morpholine
Image of 3-<WBR>Chloro-<WBR>4-<WBR>morpholin-<WBR>4-<WBR>yl-<WBR>1,2,5-<WBR>thiadiazole
CAS Number:
Empirical Formula (Hill Notation):
 C6H8ClN3OS
Molecular Weight:
 205.67
 EC Number:
 MDL number:
 NACRES:
 NA.22
 PubChem Substance ID
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More