• USA Home
  • Search Results

  • 3chlorobenzoylchloride1750161846211

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

3-Chlorobenzoyl chloride

1 Products
Image of 3-Chlorobenzoyl chloride
CAS Number:
Linear Formula:
 ClC6H4COCl
Molecular Weight:
 175.01
 Beilstein/REAXYS Number:
 386256
 EC Number:
 MDL number:
 PubChem Substance ID:
Popular Documents:  FTNMR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More