• USA Home
  • Search Results

  • 3chlorobenzylbromide2054876680311

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

3-Chlorobenzyl bromide

1 Products
Synonym:   α-Bromo-3-chlorotoluene
Image of 3-Chlorobenzyl bromide
CAS Number:
Linear Formula:
 ClC6H4CH2Br
Molecular Weight:
 205.48
 Beilstein/REAXYS Number:
 636504
 EC Number:
 MDL number:
 NACRES:
 NA.22
 PubChem Substance ID:
Popular Documents:  FTNMR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More