• USA Home
  • Search Results

  • 3chlorophenylhydrazinehydrochloride17905231223411

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

3-Chlorophenylhydrazine hydrochloride

1 Products
Image of 3-<WBR>Chlorophenylhydrazine hydrochloride
CAS Number:
Linear Formula:
 ClC6H4NHNH2 · HCl
Molecular Weight:
 179.05
 Beilstein Registry Number:
 3565828
 EC Number:
 MDL number:
 PubChem Substance ID
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Assay
Show More