• USA Home
  • Search Results

  • 3chloropropanesulfonylchloride17705163382511

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

3-Chloropropanesulfonyl chloride

1 Products
Image of 3-<WBR>Chloropropanesulfonyl chloride
CAS Number:
Linear Formula:
 Cl(CH2)3SO2Cl
Molecular Weight:
 177.05
 Beilstein/REAXYS Number:
 1754119
 EC Number:
 MDL number:
 PubChem Substance ID:
Popular Documents:  FTNMR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More