• USA Home
  • Search Results

  • 3chloropropionicacid1085210794811

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

3-Chloropropionic acid

1 Products
Image of 3-Chloropropionic acid
CAS Number:
Linear Formula:
 ClCH2CH2COOH
Molecular Weight:
 108.52
 Beilstein/REAXYS Number:
 1098495
 EC Number:
 MDL number:
 NACRES:
 NA.22
 PubChem Substance ID:
Popular Documents:  FTNMR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More