• USA Home
  • Search Results

  • 3decanone1562792880311

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

3-Decanone

1 Products
Synonym:   Ethyl heptyl ketone
Image of 3-Decanone
CAS Number:
Linear Formula:
 CH3(CH2)6COC2H5
Molecular Weight:
 156.27
 Beilstein/REAXYS Number:
 1748021
 EC Number:
 FEMA Number:
 3966
 Flavis number:
 7.151
 MDL number:
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)  |  FTNMR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Assay
  Organoleptic property
  Biological Source
  Special Grade
Show More

Product #
Description
W396605 3-Decanone, ≥97%, FG,