• USA Home
  • Search Results

  • 3dimethylamino1propyne8313722338311

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

3-Dimethylamino-1-propyne

1 Products
Synonym:   1-Dimethylamino-2-propyne, N,N-Dimethyl-2-propynylamine, N,N-Dimethylpropargylamine
Image of 3-<WBR>Dimethylamino-<WBR>1-<WBR>propyne
CAS Number:
Linear Formula:
 (CH3)2NCH2C≡CH
Molecular Weight:
 83.13
 Beilstein/REAXYS Number:
 1071222
 EC Number:
 MDL number:
 NACRES:
 NA.22
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)  |  FTNMR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More