• USA Home
  • Search Results

  • 3dxnanoesdcntpetgcarbonnanotubereinforcedpolyethyleneterephthalateglycolcopolymer3dprintingfilament123452503859911

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

3DXNANO ESD CNT-PETG carbon nanotube reinforced polyethylene terephthalate glycol copolymer 3D printing filament

1 Products
CAS Number:
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Color
  Weight
  Composite Dimension
Show More