• USA Home
  • Search Results

  • 3epi25hydroxyvitamind2solution4126590812648711

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

3-epi-25-Hydroxyvitamin D2 solution

2 Products
Image of 3-<I>epi</I>-25-Hydroxyvitamin D<SUB>2</SUB> solution
CAS Number:
Empirical Formula (Hill Notation):
 C28H44O2
Molecular Weight:
 412.65
 EC Number:
 MDL number:
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Assay
  Physical Form
  Titertest
  Concentration
  Color
Show More