• USA Home
  • Search Results

  • 3epi25hydroxyvitamind3solution400647380905911

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

3-epi-25-Hydroxyvitamin D3 solution

3 Products
Image of 3-<I>epi</I>-25-Hydroxyvitamin D<SUB>3</SUB> solution
CAS Number:
Empirical Formula (Hill Notation):
 C27H44O2
Molecular Weight:
 400.64
 EC Number:
 MDL number:
 NACRES:
 NA.24
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Concentration
  Physical Form
  Titertest
  Color
  Assay
  Melting Point
Show More