• USA Home
  • Search Results

  • 3ethyl1pentyn3ol11217628506911

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

3-Ethyl-1-pentyn-3-ol

1 Products
Synonym:   (1-Ethyl-1-hydroxypropyl)acetylene, 3-Ethylpent-1-yn-3-ol
Image of 3-<WBR>Ethyl-<WBR>1-<WBR>pentyn-<WBR>3-<WBR>ol
CAS Number:
Linear Formula:
 CH≡CC(OH)(CH2CH3)2
Molecular Weight:
 112.17
 Beilstein/REAXYS Number:
 1740944
 EC Number:
 MDL number:
 NACRES:
 NA.22
 PubChem Substance ID
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More