• USA Home
  • Search Results

  • 3ethyl52hydroxyethyl4methylthiazoliumbromide252175401670511

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

3-Ethyl-5-(2-hydroxyethyl)-4-methylthiazolium bromide

1 Products
Image of 3-<WBR>Ethyl-<WBR>5-<WBR>(2-<WBR>hydroxyethyl)<WBR>-<WBR>4-<WBR>methylthiazolium bromide
CAS Number:
Empirical Formula (Hill Notation):
 C8H14BrNOS
Molecular Weight:
 252.17
 Beilstein/REAXYS Number:
 4165775
 EC Number:
 MDL number:
 NACRES:
 NA.22
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)  |  FTNMR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More