• USA Home
  • Search Results

  • 3ethylpyridine1071553678711

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

3-Ethylpyridine

2 Products
Image of 3-Ethylpyridine
CAS Number:
Empirical Formula (Hill Notation):
 C7H9N
Molecular Weight:
 107.15
 Beilstein Registry Number:
 106479
 EC Number:
 MDL number:
 PubChem Substance ID:
Popular Documents:  FTNMR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Special Grade
  Solubility
  Assay
Show More