• USA Home
  • Search Results

  • 3ethynylaaatrifluorotoluene1701370528211

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

3-Ethynyl-α,α,α-trifluorotoluene

1 Products
Image of 3-Ethynyl-<I>α,α,α</I>-trifluorotoluene
CAS Number:
Linear Formula:
 HC≡CC6H4CF3
Molecular Weight:
 170.13
 MDL number:
 PubChem Substance ID
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More