• USA Home
  • Search Results

  • 3fluorobenzaldehyde1241145648411

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

3-Fluorobenzaldehyde

1 Products
Image of 3-Fluorobenzaldehyde
CAS Number:
Linear Formula:
 FC6H4CHO
Molecular Weight:
 124.11
 Beilstein/REAXYS Number:
 970178
 EC Number:
 MDL number:
 NACRES:
 NA.22
 PubChem Substance ID:
Popular Documents:  FTNMR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More

Product #
Description
F5005 3-Fluorobenzaldehyde, 97%,