• USA Home
  • Search Results

  • 3fluorobenzylalcohol1261345647311

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

3-Fluorobenzyl alcohol

1 Products
Image of 3-Fluorobenzyl alcohol
CAS Number:
Linear Formula:
 FC6H4CH2OH
Molecular Weight:
 126.13
 Beilstein/REAXYS Number:
 2242511
 EC Number:
 MDL number:
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)  |  FTNMR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More