• USA Home
  • Search Results

  • 3formylphenylboronicacid149948719916411

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

3-Formylphenylboronic acid

1 Products
Synonym:   (3-Formylbenzene)boronic acid, 3-(Dihydroxyboryl)benzaldehyde, 3-Boronobenzaldehyde, m-formyl-benzeneboronic acid, m-Formylphenylboronic acid
Image of 3-<WBR>Formylphenylboronic acid
CAS Number:
Linear Formula:
 HCOC6H4B(OH)2
Molecular Weight:
 149.94
 Beilstein/REAXYS Number:
 3030769
 MDL number:
 NACRES:
 NA.22
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)  |  FTIR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More