• USA Home
  • Search Results

  • 3furancarboxaldehyde960849860211

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

3-Furancarboxaldehyde

1 Products
Synonym:   3-Furaldehyde
Image of 3-<WBR>Furancarboxaldehyde
CAS Number:
Empirical Formula (Hill Notation):
 C5H4O2
Molecular Weight:
 96.08
 Beilstein/REAXYS Number:
 105852
 MDL number:
 NACRES:
 NA.22
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)  |  FTNMR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Assay
Show More