• USA Home
  • Search Results

  • 3glycidyloxypropyltrimethoxysilane23634253083811

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

(3-Glycidyloxypropyl)trimethoxysilane

1 Products
Synonym:   3-(2,3-Epoxypropoxy)propyltrimethoxysilane, GLYMO, Glycidyl 3-(trimethoxysilyl)propyl ether
Image of (3-<WBR>Glycidyloxypropyl)<WBR>trimethoxysilane
CAS Number:
Empirical Formula (Hill Notation):
 C9H20O5Si
Molecular Weight:
 236.34
 Beilstein/REAXYS Number:
 4308125
 EC Number:
 MDL number:
 NACRES:
 NA.23
 PubChem Substance ID:
Popular Documents:  FTNMR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More