• USA Home
  • Search Results

  • 3hexanol1021762337011

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

3-Hexanol

3 Products
Synonym:   Ethyl propyl carbinol
Image of 3-Hexanol
CAS Number:
Linear Formula:
 CH3CH2CH2CH(OH)CH2CH3
Molecular Weight:
 102.17
 Beilstein/REAXYS Number:
 1718964
 EC Number:
 MDL number:
 NACRES:
 NA.24
 PubChem Substance ID
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Assay
Show More