• USA Home
  • Search Results

  • 3hexyne821492849411

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

3-Hexyne

1 Products
Image of 3-Hexyne
CAS Number:
Linear Formula:
 C2H5C≡CC2H5
Molecular Weight:
 82.14
 Beilstein/REAXYS Number:
 1731158
 EC Number:
 MDL number:
 NACRES:
 NA.22
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)  |  FTNMR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More

Product #
Description
306894 3-Hexyne, 99%,