• USA Home
  • Search Results

  • 3hydroxy4methoxybenzylalcohol15416438306611

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

3-Hydroxy-4-methoxybenzyl alcohol

1 Products
Synonym:   Isovanillyl alcohol
Image of 3-<WBR>Hydroxy-<WBR>4-<WBR>methoxybenzyl alcohol
CAS Number:
Linear Formula:
 HOC6H3(OCH3)CH2OH
Molecular Weight:
 154.16
 Beilstein/REAXYS Number:
 1867860
 EC Number:
 MDL number:
 NACRES:
 NA.22
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)  |  FTNMR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More