• USA Home
  • Search Results

  • 3hydroxypicolinicacid1391187424811

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

3-Hydroxypicolinic acid

1 Products
Synonym:   3-Hydroxypyridine-2-carboxylic acid, 3-HPA
Image of 3-Hydroxypicolinic acid
CAS Number:
Empirical Formula (Hill Notation):
 C6H5NO3
Molecular Weight:
 139.11
 Beilstein/REAXYS Number:
 118954
 EC Number:
 MDL number:
 NACRES:
 NA.21
 PubChem Substance ID
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Assay
Show More