• USA Home
  • Search Results

  • 3hydroxypyridine951010900211

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

3-Hydroxypyridine

1 Products
Synonym:   3-Pyridinol, 3-Pyridone
Image of 3-Hydroxypyridine
CAS Number:
Empirical Formula (Hill Notation):
 C5H5NO
Molecular Weight:
 95.10
 Beilstein/REAXYS Number:
 105699
 EC Number:
 MDL number:
 PubChem Substance ID:
Popular Documents:  FTNMR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Assay
Show More

Product #
Description
H57009 3-Hydroxypyridine, 98%,