• USA Home
  • Search Results

  • 3hydroxytyrosol154161059760111

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

3-Hydroxytyrosol

2 Products
Synonym:   2-(3,4-Dihydroxyphenyl)ethanol, 3,4-Dihydroxyphenethyl alcohol, DOPET, Homoprotocatechuyl alcohol
Image of 3-Hydroxytyrosol
CAS Number:
Empirical Formula (Hill Notation):
 C8H10O3
Molecular Weight:
 154.16
 Beilstein/REAXYS Number:
 2208118
 MDL number:
 NACRES:
 NA.25
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Assay
Show More