• USA Home
  • Search Results

  • 3iodo2propynylnbutylcarbamate281095540653611

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

3-Iodo-2-propynyl N-butylcarbamate

2 Products
Image of 3-Iodo-2-propynyl <I>N</I>-butylcarbamate
CAS Number:
Linear Formula:
 CH3(CH2)3NHCO2CH2C≡Cl
Molecular Weight:
 281.09
 EC Number:
 MDL number:
 NACRES:
 NA.22
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Assay
  Application(s)
Show More