• USA Home
  • Search Results

  • 3iodobenzoicacid2480261851911

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

3-Iodobenzoic acid

1 Products
Image of 3-Iodobenzoic acid
CAS Number:
Linear Formula:
 IC6H4CO2H
Molecular Weight:
 248.02
 Beilstein/REAXYS Number:
 971088
 EC Number:
 MDL number:
 NACRES:
 NA.22
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)  |  FTNMR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Assay
Show More

Product #
Description
138584 3-Iodobenzoic acid, 98%,