• USA Home
  • Search Results

  • 3mercaptopropionicacid1061410796011

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

3-Mercaptopropionic acid

3 Products
Synonym:   3-sulfanylpropanoic acid
Image of 3-Mercaptopropionic acid
CAS Number:
Linear Formula:
 HSCH2CH2CO2H
Molecular Weight:
 106.14
 Beilstein/REAXYS Number:
 773807
 EC Number:
 MDL number:
 NACRES:
 NA.23
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)  |  FTNMR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Assay
Show More