• USA Home
  • Search Results

  • 3methoxypropylamine8914533273011

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

3-Methoxypropylamine

2 Products
Synonym:   1-Amino-3-methoxypropane
Image of 3-Methoxypropylamine
CAS Number:
Linear Formula:
 CH3O(CH2)3NH2
Molecular Weight:
 89.14
 Beilstein/REAXYS Number:
 878144
 EC Number:
 MDL number:
 NACRES:
 NA.22
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)  |  FTNMR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Assay
Show More