• USA Home
  • Search Results

  • 3methyl1butanol881512351311

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

3-Methyl-1-butanol

6 Products
Synonym:   Isoamyl alcohol, Isopentyl alcohol
Image of 3-Methyl-1-butanol
CAS Number:
Linear Formula:
 (CH3)2CHCH2CH2OH
Molecular Weight:
 88.15
 Beilstein/REAXYS Number:
 1718835
 EC Number:
 MDL number:
 NACRES:
 NA.21
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Special Grade
  Restriction
  Product Line
  Application(s)
  pH Value
  Physical Form
  Assay
Show More