• USA Home
  • Search Results

  • 3methyl1butene701356345111

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

3-Methyl-1-butene

1 Products
Image of 3-Methyl-1-butene
CAS Number:
Linear Formula:
 (CH3)2CHCH=CH2
Molecular Weight:
 70.13
 Beilstein/REAXYS Number:
 505980
 EC Number:
 MDL number:
 NACRES:
 NA.22
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)  |  FTNMR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Assay
  Application(s)
Show More

Product #
Description
257931 3-Methyl-1-butene, 95%,