• USA Home
  • Search Results

  • 3methyl1penten4yn3ol9613323069111

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

3-Methyl-1-penten-4-yn-3-ol

1 Products
Synonym:   Ethynyl methyl vinyl carbinol
Image of 3-<WBR>Methyl-<WBR>1-<WBR>penten-<WBR>4-<WBR>yn-<WBR>3-<WBR>ol
CAS Number:
Linear Formula:
 HC≡CC(CH3)(OH)CH=CH2
Molecular Weight:
 96.13
 EC Number:
 MDL number:
 NACRES:
 NA.22
 PubChem Substance ID
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More