• USA Home
  • Search Results

  • 3methyl1pentyn3aminehydrochloride1336210857532211

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

3-Methyl-1-pentyn-3-amine hydrochloride

1 Products
Image of 3-<WBR>Methyl-<WBR>1-<WBR>pentyn-<WBR>3-<WBR>amine hydrochloride
CAS Number:
Empirical Formula (Hill Notation):
 C6H11N · HCl
Molecular Weight:
 133.62
 MDL number:
 NACRES:
 NA.22
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More