• USA Home
  • Search Results

  • 3methyl1pentyn3ol98147775811

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

3-Methyl-1-pentyn-3-ol

1 Products
Synonym:   Ethyl ethynyl methyl carbinol, Meparfynol
Image of 3-<WBR>Methyl-<WBR>1-<WBR>pentyn-<WBR>3-<WBR>ol
CAS Number:
Linear Formula:
 CH≡CC(OH)(CH3)CH2CH3
Molecular Weight:
 98.14
 Beilstein/REAXYS Number:
 969340
 EC Number:
 MDL number:
 NACRES:
 NA.22
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)  |  FTNMR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Solubility
Show More