• USA Home
  • Search Results

  • 3methyl2benzothiazolinonehydrazonehydrochloridehydrate21570anhydrousbasis14902215111

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

3-Methyl-2-benzothiazolinone hydrazone hydrochloride hydrate

1 Products
Synonym:   MBTH hydrochloride hydrate
Image of 3-<WBR>Methyl-<WBR>2-<WBR>benzothiazolinone hydrazone hydrochloride hydrate
CAS Number:
Empirical Formula (Hill Notation):
 C8H9N3S · HCl · xH2O
Molecular Weight:
 215.70 (anhydrous basis)
 EC Number:
 MDL number:
 NACRES:
 NA.22
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)  |  FTIR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More