• USA Home
  • Search Results

  • 3methylbutanol881512351311

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

3-Methylbutanol

4 Products
Synonym:   3-Methyl-1-butanol, Isoamyl alcohol, Isopentyl alcohol
Image of 3-Methylbutanol
CAS Number:
Linear Formula:
 (CH3)2CHCH2CH2OH
Molecular Weight:
 88.15
 Beilstein Registry Number:
 1718835
 EC Number:
 MDL number:
 PubChem Substance ID:
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Assay
  UV-Absorption Values
  Titertest
  Product Line
  Physical Form
  Refractive Index
  Special Grade
Show More