• USA Home
  • Search Results

  • 3methylpyrazole8210145358311

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

3-Methylpyrazole

1 Products
Image of 3-Methylpyrazole
CAS Number:
Empirical Formula (Hill Notation):
 C4H6N2
Molecular Weight:
 82.10
 Beilstein/REAXYS Number:
 1454
 EC Number:
 MDL number:
 PubChem Substance ID:
Popular Documents:  FTIR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Assay
Show More

Product #
Description
M75802 3-Methylpyrazole, 97%,